missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-fluorobenzene, 98+%

Product Code. 11424717
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424717 25 g 25g
11434717 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424717 Supplier Thermo Scientific Alfa Aesar Supplier No. B21209.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 348-51-6 | C6H4ClF | 130.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 348-51-6
Molecular Formula C6H4ClF
Molecular Weight (g/mol) 130.55
MDL Number MFCD00000533
InChI Key ZCJAYDKWZAWMPR-UHFFFAOYSA-N
Synonym 2-chlorofluorobenzene, o-chlorofluorobenzene, benzene, 1-chloro-2-fluoro, o-fluorochlorobenzene, 1-fluoro-2-chlorobenzene, 1-chloro-2-fluoro-benzene, benzene, chlorofluoro, 2-chloro-1-fluorobenzene, chlorofluorobenzene, 2-fluorochlorobenzene
PubChem CID 9583
IUPAC Name 1-chloro-2-fluorobenzene
SMILES FC1=CC=CC=C1Cl

Specifications

Melting Point -43°C
Density 1.242
Boiling Point 137°C to 138°C
Flash Point 31°C (87°F)
Refractive Index 1.501
Quantity 25 g
UN Number UN1993
Beilstein 1855301
Formula Weight 130.55
Percent Purity ≥98%
Chemical Name or Material 1-Chloro-2-fluorobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.