missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopropyl-6,7-difluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid, Mikromol™

Product Code. 30051608
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051608 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051608 Supplier Mikromol Supplier No. MM0885.170025
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-Cyclopropyl-6,7-difluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid
TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid
CAS 93107-30-3
Analyte or Component Names 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid
Shipping Condition Room Temperature
Molecular Formula C13 H9 F2 N O3
Certifications/Compliance ISO 9001
Synonym 3-Quinolinecarboxylic acid, 1-cyclopropyl-6,7-difluoro-1,4-dihydro-4-oxo-, 1-cyclopropyl-6,7-difluoro-4-oxo-quinoline-3-carboxylic acid, 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid
InChI Formula InChI=1S/C13H9F2NO3/c14-9-3-7-11(4-10(9)15)16(6-1-2-6)5-8(12(7)17)13(18)19/h3-6H,1-2H2,(H,18,19)
SMILES OC(=O)C1=CN(C2CC2)c3cc(F)c(F)cc3C1=O
IUPAC Name 1-cyclopropyl-6,7-difluoro-4-oxoquinoline-3-carboxylic acid
Molecular Weight (g/mol) 265.2123
Quantity 25 mg
Formula Weight 265.055 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.