missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-4-iodobenzene, 99%

Product Code. 11444064
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444064 250 g 250g
11424064 10 g 10g
11434064 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444064 Supplier Thermo Scientific Alfa Aesar Supplier No. A12008.30
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 352-34-1 | C6H4FI | 222.00 g/mol

4-Fluoroiodobenzene was used in the preparation of fluorotriphenylene derivatives. 4-[F] Fluoroiodobenzene is a versatile building block in 18F radiochemistry and is used in various transition metal-mediated C-C and C-N cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoroiodobenzene was used in the preparation of fluorotriphenylene derivatives. 4-[F] Fluoroiodobenzene is a versatile building block in 18F radiochemistry and is used in various transition metal-mediated C-C and C-N cross-coupling reactions.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool dry place. Keep container tightly sealed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352-34-1
Molecular Formula C6H4FI
Molecular Weight (g/mol) 222.00
MDL Number MFCD00001052
InChI Key KGNQDBQYEBMPFZ-UHFFFAOYSA-N
Synonym 4-fluoroiodobenzene, p-fluoroiodobenzene, benzene, 1-fluoro-4-iodo, 4-iodofluorobenzene, p-iodofluorobenzene, 4-fluoro-1-iodobenzene, 4-fluoro-iodobenzene, 1-fluoro-4-iodo-benzene, 1-iodo-4-fluorobenzene, 4-fluroiodobenzene
PubChem CID 9605
IUPAC Name 1-fluoro-4-iodobenzene
SMILES FC1=CC=C(I)C=C1

Specifications

Melting Point -20°C
Density 1.925
Boiling Point 182°C to 184°C
Flash Point 68°C (154°F)
Refractive Index 1.583
Quantity 250 g
Beilstein 1853970
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 222
Percent Purity 99%
Chemical Name or Material 1-Fluoro-4-iodobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.