missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxypyrene, 99+%

Product Code. 10286380 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
50mg
This item is not returnable. View return policy

Product Code. 10286380

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5315-79-7 | C16H10O | 218.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5315-79-7
Molecular Formula C16H10O
Molecular Weight (g/mol) 218.25
MDL Number MFCD00044543
InChI Key BIJNHUAPTJVVNQ-UHFFFAOYSA-N
Synonym 1-hydroxypyrene, 1-pyrenol, 3-hydroxypyrene, pyrenol, unii-n2h6o5v707, hydroxypyrene, 1-hydroxy-pyrene, 3-pyrenol, 1-pyrenol 8ci
PubChem CID 21387
ChEBI CHEBI:34093
IUPAC Name pyren-1-ol
SMILES C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 176°C to 178°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in most common organic solvents
Formula Weight 218.25
Percent Purity 99+%
Physical Form Powder or Fine Needles
Chemical Name or Material 1-Hydroxypyrene, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Hydroxypyrene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.