missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthol, Mikromol™

Product Code. 30050491
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050491 25 mg 25mg
18080560 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050491 Supplier Mikromol Supplier No. MM0043.040025
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-Naphthol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Naphthol
CAS 90-15-3
Analyte or Component Names 1-Naphthol
Shipping Condition Room Temperature
Molecular Formula C10 H8 O
Certifications/Compliance ISO 17025
Synonym Naphthalen-1-ol, 1-Naphthol, Duloxetine Hydrochloride Imp. D (Pharmeuropa), Duloxetine Hydrochloride Imp. D (EP), Duloxetine Imp. D (EP), 1-Naphthalenol, 1-Naphthol (8CI), 1-Hydroxynaphthalene, 1-Naphthyl alcohol, BASF Ursol ERN, C.I. 76605, C.I. Oxidation Base 33, Durafur Developer D, Fouramine ERN, Fourrine 99, Fourrine ERN, Furro ER, NSC 9586, Nako TRB, Naphthol-1, Naphthyl-1-ol, Tertral ERN, Ursol ERN, Zoba ERN, alpha-Hydroxynaphthalene, alpha-Naphthol, alpha-Naphthyl alcohol, Duloxetine Hydrochloride Impurity D, Duloxetine Impurity D
InChI Formula InChI=1S/C10H8O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7,11H
SMILES Oc1cccc2ccccc12
IUPAC Name naphthalen-1-ol
Molecular Weight (g/mol) 144.17
Quantity 25 mg
Formula Weight 144.0575 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.