missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylpiperazine Hydrochloride, Mikromol™

Product Code. 30050665
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050665 25 mg 25mg
18031620 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050665 Supplier Mikromol Supplier No. MM1246.020025
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-Phenylpiperazine Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Phenylpiperazine Hydrochloride
CAS 2210-93-7
Analyte or Component Names 1-Phenylpiperazine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C10 H14 N2 . Cl H
Certifications/Compliance ISO 9001
Synonym Levodropropizine Imp. B (EP), 1-Phenylpiperazine Hydrochloride, Levodropropizine Imp. B (EP) as Hydrochloride, Levodropropizine Impurity B as Hydrochloride
InChI Formula InChI=1S/C10H14N2.ClH/c1-2-4-10(5-3-1)12-8-6-11-7-9-12;/h1-5,11H,6-9H2;1H
SMILES Cl.C1CN(CCN1)c2ccccc2
IUPAC Name 1-phenylpiperazine;hydrochloride
Molecular Weight (g/mol) 198.69
Quantity 25 mg
Formula Weight 198.0924 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.