missing translation for 'onlineSavingsMsg'
Learn More

1-(Phenylsulfonyl)-1H-indol-3-ylboronic acid, 97%, May contain varying amounts of anhydri, Thermo Scientific™

Product Code. 10232352
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10232352 250 mg Amber Glass Bottle 250mg
10242352 1 g Amber Glass Bottle 1g
10386794 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10232352 Supplier Thermo Scientific Maybridge Supplier No. CC03112CB
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 129271-98-3
Molecular Formula C14H12BNO4S
Molecular Weight (g/mol) 301.12
MDL Number MFCD02681892
InChI Key YKTZLHLBQGCFQX-UHFFFAOYSA-N
Synonym 1-phenylsulfonyl-3-indoleboronic acid, 1-phenylsulfonyl-1h-indol-3-ylboronic acid, 1-phenylsulfonyl-1h-indol-3-yl boronic acid, 1-benzenesulfonyl indol-3-yl boronic acid, 1-phenylsulfonylindole-3-boronic acid, 1-phenylsulphonyl-1h-indole-3-boronic acid, 1-phenylsulfonyl-3-indolylboronic acid, boronic acid, b-1-phenylsulfonyl-1h-indol-3-yl, 1-benzenesulfonyl indol-3-ylboronic acid, 1-benzenesulfonyl-1h-indol-3-yl boranediol
PubChem CID 2776217
IUPAC Name [1-(benzenesulfonyl)indol-3-yl]boronic acid
SMILES OB(O)C1=CN(C2=CC=CC=C12)S(=O)(=O)C1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 301.13
Percent Purity 97%
Chemical Name or Material 1-(Phenylsulfonyl)-1H-indol-3-ylboronic acid, May contain varying amounts of anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.