missing translation for 'onlineSavingsMsg'
Learn More

10-[2-[(2RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulfonyl)-10H-phenothiazine 5,5-Dioxide (Sulforidazine 5-Sulfone), Mikromol™

Product Code. 30051253
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051253 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051253 Supplier Mikromol Supplier No. MM0116.110025
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

10-[2-[(2RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulfonyl)-10H-phenothiazine 5,5-Dioxide (Sulforidazine 5-Sulfone)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulforidazine 5-Sulfone
CAS 100574-22-9
Analyte or Component Names Sulforidazine 5-Sulfone
Shipping Condition Room Temperature
Molecular Formula C21 H26 N2 O4 S2
Certifications/Compliance ISO 9001
Synonym 10-[2-(1-Methyl-2-piperidinyl)ethyl]-2-(methylsulfonyl)-10H-phenothiazine 5,5-Dioxide, Sulforidazine 5-Sulfone, Thioridazine Hydrochloride Imp. A (EP), Thioridazine Imp. A (EP), Thioridazine Hydrochloride Impurity A, Thioridazine Impurity A
InChI Formula InChI=1S/C21H26N2O4S2/c1-22-13-6-5-7-16(22)12-14-23-18-8-3-4-9-20(18)29(26,27)21-11-10-17(15-19(21)23)28(2,24)25/h3-4,8-11,15-16H,5-7,12-14H2,1-2H3
SMILES CN1CCCCC1CCN2c3ccccc3S(=O)(=O)c4ccc(cc24)S(=O)(=O)C
IUPAC Name 10-[2-(1-methylpiperidin-2-yl)ethyl]-2-methylsulfonylphenothiazine 5,5-dioxide
Molecular Weight (g/mol) 434.57
Quantity 25 mg
Formula Weight 434.1334 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.