missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avanafil, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Avanafil
Specifications
Specifications
| Chemical Name or Material | Avanafil |
| CAS | 330784-47-9 |
| Analyte or Component Names | Avanafil |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H26 Cl N7 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 5-Pyrimidinecarboxamide, 4-[[(3-chloro-4-methoxyphenyl)methyl]amino]-2-[(2S)-2-(hydroxymethyl)-1-pyrrolidinyl]-N-(2-pyrimidinylmethyl)-, (S)-2-(2-Hydroxymethyl-1-pyrrolidinyl)-4-(3-chloro-4-methoxybenzylamino)-5-[(2-pyrimidinylmethyl)carbamoyl]pyrimidine, Avanafil, TA 1790 |
| InChI Formula | InChI=1S/C23H26ClN7O3/c1-34-19-6-5-15(10-18(19)24)11-27-21-17(22(33)28-13-20-25-7-3-8-26-20)12-29-23(30-21)31-9-2-4-16(31)14-32/h3,5-8,10,12,16,32H,2,4,9,11,13-14H2,1H3,(H,28,33)(H,27,29,30)/t16-/m0/s1 |
| SMILES | COc1ccc(CNc2nc(ncc2C(=O)NCc3ncccn3)N4CCC[C@H]4CO)cc1Cl |
| IUPAC Name | 4-[(3-chloro-4-methoxyphenyl)methylamino]-2-[(2S)-2-(hydroxymethyl)pyrrolidin-1-yl]-N-(pyrimidin-2-ylmethyl)pyrimidine-5-carboxamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction