missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone 17-heptanoate, Lipomed
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Testosterone 17-heptanoate
Specifications
Specifications
| Chemical Name or Material | Testosterone enanthate |
| CAS | 315-37-7 |
| Analyte or Component Names | Testosterone enanthate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H40 O3 |
| Synonym | Testosterone Isocaproate Imp. F (EP), Testosterone Enantate, Androst-4-en-3-one, 17-[(1-oxoheptyl)oxy]-, (17 beta)-, Testosterone, heptanoate (6 CI,8 CI), (17 beta)-17-[(1-Oxoheptyl)oxy]androst-4-en-3-one, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate, 4-Androsten-3-one 17 beta-enanthate |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CCCCCCC(=O)O[C@H]1 CC[C@H]2 [C@@H]3 CCC4=CC(=O)CC[C@]4(C)[C@H]3 CC[C@]12 C |
| IUPAC Name | [(8 R,9 S,10 R,13 S,14 S,17 S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] heptanoate |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction