missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NKG2D/CD314 Polyclonal specifically detects NKG2D/CD314 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
Specifications
| Dilution | Western Blot 0.1 - 0.5 μg/mL, Flow Cytometry 10 μg/mL, Peptide ELISA Detection limit 1:64000 |
| Gene Alias | CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence |
| Host Species | Goat |
| Immunogen | Peptide with sequence C-KVYSKEDQDLLK corresponding to internal region according to NP_031386.2. |
| Quantity | 0.1 mg |
| Regulatory Status | RUO |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?