missing translation for 'onlineSavingsMsg'
Learn More

Connexin 32/GJB1 Antibody (GJB1/1753), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18755051 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18755051 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18755051 Supplier Novus Biologicals Supplier No. NBP254359AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Connexin 32/GJB1 Monoclonal antibody specifically detects Connexin 32/GJB1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 32/GJB1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone GJB1/1753
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias CMTX, Connexin-32, Cx32, CX32beta 1, 32kD (connexin 32, Charcot-Marie-Toothneuropathy, X-linked), gap junction beta-1 protein, gap junction protein, beta 1, 32kDa, gap junction protein, beta 1, 32kDa (connexin 32), gap junction protein, beta 1, 32kDa (connexin 32, Charcot-Marie-Toothneuropathy, X-linked)
Host Species Mouse
Immunogen Recombinant human Connexin 32/GJB1 protein (Uniprot: P08034)
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2705
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.