missing translation for 'onlineSavingsMsg'
Learn More

GluR1, p Ser845 Antibody (S03-7G4) - AMPA Subtype, Novus Biologicals™

Product Code. 18685034 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18685034 100 μg 100µL
18675744 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18685034 Supplier Novus Biologicals Supplier No. NBP319943100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GluR1 Monoclonal antibody specifically detects GluR1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR1
Applications Western Blot
Classification Monoclonal
Clone S03-7G4
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias AMPA 1, AMPA-selective glutamate receptor 1, GluA1, GLUH1, GluR-1, GLUR1gluR-A, GluR-A, GLURA, GluR-K1, glutamate receptor 1, Glutamate receptor ionotropic, AMPA 1, glutamate receptor, ionotropic, AMPA 1, HBGR1, MGC133252
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Ser845 of human AMPA Receptor 1 (GluR1) (Uniprot # P42261)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2890
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.