missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NRIP Polyclonal specifically detects NRIP in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).
Specifications
Specifications
| Applications | Western Blot |
| Classification | Polyclonal |
| Quantity | 100 μL |
| Isotype | IgG |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?