missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deanol Acetamidobenzoate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Deanol Acetamidobenzoate
Specifications
Specifications
| Chemical Name or Material | Deanol Acetamidobenzoate |
| CAS | 3635-74-3 |
| Analyte or Component Names | Deanol Acetamidobenzoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H9 N O3 . C4 H11 N O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzoic acid, p-acetamido-, compd. with 2-(dimethylamino)ethanol (1:1), Benzoic acid, p-acetamido-, compd. with 2-(dimethylamino)ethanol, Ethanol, 2-(dimethylamino)-, 4-(acetylamino)benzoate (salt), Ethanol, 2-(dimethylamino)-, compd. with p-acetamidobenzoic acid (1:1), Ethanol, 2-(dimethylamino)-, p-acetamidobenzoate (salt), 2-(Dimethylamino)ethanol p-acetylaminobenzoate, 2-Dimethylaminoethanol p-acetamidobenzoate, Deaner, Deanol Acetamidobenzoate, Diforene, NSC 97399, Pabenol |
| InChI Formula | InChI=1 S/C9H9NO3.C4H11NO/c1-6(11)10-8-4-2-7(3-5-8)9(12)13;1-5(2)3-4-6/h2-5 H,1H3,(H,10,11)(H,12,13);6 H,3-4H2,1-2H3 |
| SMILES | CN(C)CCO.CC(=O)Nc1ccc(cc1)C(=O)O |
| IUPAC Name | 4-acetamidobenzoic acid;2-(dimethylamino)ethanol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction