missing translation for 'onlineSavingsMsg'
Learn More

Doxylamine.succinate, Lipomed

Product Code. 30464237
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30464237

Brand: Lipomed LPMDXL1220SU10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Doxylamine.succinate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Doxilamine succinate
CAS 562-10-7
Analyte or Component Names Doxilamine succinate
Shipping Condition Room Temperature
Molecular Formula C17 H22 N2 O . C4 H6 O4
Synonym Doxylamine Hydrogen Succinate, Butanedioic acid, compd. with N,N-dimethyl-2-[1-phenyl-1-(2-pyridinyl)ethoxy]ethanamine (1:1), Ethanamine, N,N-dimethyl-2-[1-phenyl-1-(2-pyridinyl)ethoxy]-, butanedioate (1:1) (9 CI), Alsodorm, Decamium, Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI)
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CN(C)CCOC(C)(c1ccccc1)c2ccccn2.OC(=O)CCC(=O)O
IUPAC Name butanedioic acid;N,N-dimethyl-2-(1-phenyl-1-pyridin-2-ylethoxy)ethanamine
Industry Type Pharmaceutical
Molecular Weight (g/mol) 388.46
Quantity 10 mg
Formula Weight 388.1998 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.