missing translation for 'onlineSavingsMsg'
Learn More

Metenolone enanthate, Lipomed

Product Code. 30457988
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30457988

Brand: Lipomed LPMMTN181810

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Metenolone enanthate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Primobolan
CAS 303-42-4
Analyte or Component Names Primobolan
Shipping Condition Room Temperature
Molecular Formula C27 H42 O3
Synonym Metenolone enanthate, Methenolone 17-enanthate, Methenolone enanthate, NSC 64967, Primobolan Depot, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate, 17 beta-Hydroxy-1-methyl-5 alpha-androst-1-en-3-one heptanoate
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CCCCCCC(=O)O[C@H]1 CC[C@H]2 [C@@H]3 CC[C@H]4 CC(=O)C=C(C)[C@]4(C)[C@H]3 CC[C@]12 C
IUPAC Name [(5 S,8 R,9 S,10 S,13 S,14 S,17 S)-1,10,13-trimethyl-3-oxo-4,5,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] heptanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 414.62
Quantity 10 mg
Formula Weight 414.3134 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.