missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mianserin.HCl, Lipomed
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Mianserin.HCl
Specifications
Specifications
| Chemical Name or Material | Mianserin Hydrochloride |
| CAS | 21535-47-7 |
| Analyte or Component Names | Mianserin Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H20 N2 . Cl H |
| Synonym | Mianserin Hydrochloride, Dibenzo[c,f]pyrazino[1,2-a]azepine, 1,2,3,4,10,14 b-hexahydro-2-methyl-, hydrochloride (1:1), Dibenzo[c,f]pyrazino[1,2-a]azepine, 1,2,3,4,10,14 b-hexahydro-2-methyl-, monohydrochloride, Athymil, Bolvidon, GB 94, Lantanon, Mianserin hydrochloride, Mianserine hydrochloride, NSC 292267, Norval, Org GB 94, Tetramide, Toluon, Tolvin, Tolvon |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.CN1CCN2C(C1)c3ccccc3Cc4ccccc24 |
| IUPAC Name | 1,2,3,4,10,14 b-Hexahydro-2-methyldibenzo[c,f]pyrazino[1,2-a]azepine hydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction