missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nandrolone, Lipomed
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Nandrolone
Specifications
Specifications
| Chemical Name or Material | Nandrolone |
| CAS | 434-22-0 |
| Analyte or Component Names | Nandrolone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H26 O2 |
| Synonym | 17 beta-Hydroxyestr-4-en-3-one, Nandrolone, Nandrolone Decanoate Imp. D (EP), 19-Nortestosterone, Nandrolone Decanoate Imp. D (EP), Estr-4-en-3-one, 17-hydroxy-, (17 beta)- (9 CI), d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one, d-13 beta-Methyl-17 beta-hydroxygon-4-en-3-one |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | C[C@]12 CC[C@H]3 [C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1 CC[C@@H]2 O |
| IUPAC Name | (8 R,9 S,10 R,13 S,14 S,17 S)-17-hydroxy-13-methyl-2,6,7,8,9,10,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthren-3-one |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction