missing translation for 'onlineSavingsMsg'
Learn More

11-(4-Ethylpiperazin-1-yl)dibenzo[b,f][1,4]thiazepine Dihydrochloride, Mikromol™

Product Code. 30052105
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052105 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30052105 Supplier Mikromol Supplier No. MM1130.420025
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

11-(4-Ethylpiperazin-1-yl)dibenzo[b,f][1,4]thiazepine Dihydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 11-(4-Ethylpiperazin-1-yl)dibenzo[b,f][1,4]thiazepine Dihydrochloride
CAS 2518203-12-6
Analyte or Component Names 11-(4-Ethylpiperazin-1-yl)dibenzo[b,f][1,4]thiazepine Dihydrochloride
Shipping Condition Room Temperature
Molecular Formula C19 H21 N3 S . 2 Cl H
Certifications/Compliance ISO 9001
Synonym 11-(4-Ethylpiperazin-1-yl)dibenzo[b,f][1,4]thiazepine Dihydrochloride, Quetiapine Fumarate Imp. P (EP) as Dihydrochloride, Dibenzo[b,f][1,4]thiazepine, 11-(4-ethyl-1-piperazinyl)-, hydrochloride (1:2), N-Ethyl quetiapine dihydrochloride, Quetiapine Fumarate Impurity P as Dihydrochloride, Quetiapine Impurity P as Dihydrochloride
InChI Formula InChI=1S/C19H21N3S.2ClH/c1-2-21-11-13-22(14-12-21)19-15-7-3-5-9-17(15)23-18-10-6-4-8-16(18)20-19;;/h3-10H,2,11-14H2,1H3;2*1H
SMILES Cl.Cl.CCN1CCN(CC1)C2=Nc3ccccc3Sc4ccccc24
IUPAC Name 6-(4-ethylpiperazin-1-yl)benzo[b][1,4]benzothiazepine;dihydrochloride
Molecular Weight (g/mol) 396.377
Quantity 25 mg
Formula Weight 395.099 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.