missing translation for 'onlineSavingsMsg'
Learn More

(14bRS)-1,2,3,4,10,14b-Hexahydropyrazino[2,1-a]pyrido[2,3-c][2]benzazepine (Desmethylmirtazapine), Mikromol™

Product Code. 17901366
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity unitSize
17901366 25 mg 25mg
1 options
This item is not returnable. View return policy

Product Code. 17901366

Brand: Mikromol MM1005.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(14bRS)-1,2,3,4,10,14b-Hexahydropyrazino[2,1-a]pyrido[2,3-c][2]benzazepine (Desmethylmirtazapine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Desmethylmirtazapine
CAS 61337-68-6
Analyte or Component Names N-Desmethylmirtazapine
Shipping Condition Room Temperature
Molecular Formula C16 H17 N3
Certifications/Compliance ISO 17025
Synonym N-Desmethylmirtazapine, (14bRS)-1,2,3,4,10,14b-Hexa-hydropyrazino[2,1-a]pyrido[2,3-c][2]benz-azepine, Mirtazapine Related Compound A, Mirtazapine Impurity D
InChI Formula InChI=1S/C16H17N3/c1-2-6-14-12(4-1)10-13-5-3-7-18-16(13)19-9-8-17-11-15(14)19/h1-7,15,17H,8-11H2
SMILES C1CN2C(CN1)c3ccccc3Cc4cccnc24
Molecular Weight (g/mol) 251.33
Quantity 25 mg
Formula Weight 251.1422 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.