missing translation for 'onlineSavingsMsg'
Learn More

17-beta-Estradiol, Dr. Ehrenstorfer

Product Code. 30311708
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30311708 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311708 Supplier Dr. Ehrenstorfer Supplier No. DREC13213100100MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Estradiol
CAS 50-28-2
Analyte or Component Names Estradiol
Shipping Condition +20°C
Molecular Formula C18 H24 O2
Certifications/Compliance ISO 17034
Synonym Estra-1,3,5(10)-triene-3,17-diol (17β)-, Estradiol (8CI), (17β)-Estra-1,3,5(10)-triene-3,17-diol, (+)-3,17β-Estradiol, 13β-Methyl-1,3,5(10)-gonatriene-3,17β-ol, 17β-Estradiol, 17β-Oestradiol, 3,17-Epidihydroxyestratriene, 3,17β-Dihydroxyestra-1,3,5(10)-triene, 3,17β-Estradiol, Absorlent Matrix, Aerodiol, Alora, Altrad, Aquadiol, Bardiol, Benzogynestry, Beta-estradiol, Bio-E Gel, Climaderm, Climara, Compudose, Compudose 100, Compudose 200, Compudose 365, Compudose 400, Corpagen, Dermestril, Destradiol, Dihydrofollicular hormone, Dihydrofolliculin, Dihydromenformon, Dihydrotheelin, Dihydroxyestrin, Dimenformon, Diogyn, Diogynets, Divigel, E 2, Elestrim, Encore, Epiestriol 50, Estasorb, Estra-1,3,5(10)-triene-3,17-diol, (17β)-, Estra-1,3,5(10)-triene-3,17β-diol, Estrace, Estraderm, Estraderm MX, Estraderm TTS, Estraderm TTS 100, Estraderm TTS 50, Estradiol-17β, Estradot, Estraldine, Estrapatch, Estredox, Estreva, Estring, Estring Vaginal Ring, Estroclim, Estroclim 50, Estrogel, Estrogel HBF, Estropause, Estrovite, Evamist, Evorel, FemSeven, Femanest, Femestral, Femogen, Fempatch, Follicyclin, Gelestra, Ginosedol, Gynergon, Gynodiol, Gynoestryl, Innofem, Lamdiol, Macrodiol, Menorest, Menostar, NSC 9895, Nordicol, Oesclim, Oestergon, Oestra-1,3,5(10)-triene-3,17β-diol, Oestradiol, Oestrogel, Oestroglandol, Ovahormon, Ovasterol, Ovastevol, Ovocyclin, Ovocylin, Perlatanol, Primofol, Profoliol, Profoliol B, Progynon, Progynon DH, Sandrena 1, Sprediol, Syndiol, Systen, Theelin, dihydro-, Vagifem, Vivelle, Vivelle-Dot, Zesteem, Zumenon, β-Estradiol
InChI Formula InChI=1S/C18H24O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-17,19-20H,2,4,6-9H2,1H3/t14-,15-,16+,17+,18+/m1/s1
SMILES C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CC[C@@H]2O
IUPAC Name (8R,9S,13S,14S,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
Molecular Weight (g/mol) 272.38
Quantity 100 mg
Formula Weight 272.1776 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.