missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzimidazole-2-thiol, Mikromol™

Product Code. 30052328
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052328 25 mg 25mg
18091310 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30052328 Supplier Mikromol Supplier No. MM0474.050025
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1H-Benzimidazole-2-thiol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1H-Benzimidazole-2-thiol
CAS 583-39-1
Analyte or Component Names 1H-Benzimidazole-2-thiol
Shipping Condition Room Temperature
Molecular Formula C7 H6 N2 S
Certifications/Compliance ISO 17025
Synonym Lansoprazole Imp. E (EP), Rabeprazole Sodium Imp. F (EP), Rabeprazole Imp. F (EP), Rabeprazole USP Related Compound C, Rabeprazole USP RC C, 1,3-Dihydro-2H-benzimidazole-2-thione, 2-Benzimidazolethiol (8CI), 2-Benzimidazolinethione (6CI,7CI), 1,3-Dihydro-2H-benzoimidazole-2-thione, 1H-Benzimidazole-2-thiol, 1H-Benzo[d]imidazole-2(3H)-thione, 1H-Benzo[d]imidazole-2-thiol, 2,3-Dihydro-1H-benzimidazole-2-thione, 2-Benzimidazolethione, 2-Mercapto-1H-benzimidazole, 2-Mercaptobenzimidazole, 2-Sulfhydrylbenzimidazole, 2-Thiobenzimidazole, AO-MB, ASM MB, Antage MB, Antigene MB, Antioxidant MB, Antioxidant MB/K, Benzimidazolin-2-thione, Curekind MBI/B, G 580, MB, MB/K, MBI 80, NSC 186246, NSC 21414, Nocrac MB, Nonflex MB, Permanax 21, Rhenogran MBI 80, Sandant MB, Sumilizer MB, Vulkanox MB, Vulkanox MBI, Vulkanox MG, o-Phenylenethiourea, Rabeprazole Related Compound C, Lansoprazole Impurity E, Rabeprazole Sodium Impurity F, Rabeprazole Impurity F
InChI Formula InChI=1S/C7H6N2S/c10-7-8-5-3-1-2-4-6(5)9-7/h1-4H,(H2,8,9,10)
SMILES Sc1nc2ccccc2[nH]1
IUPAC Name 1H-benzimidazole-2-thiol
Molecular Weight (g/mol) 150.2
Quantity 25 mg
Formula Weight 150.0252 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.