missing translation for 'onlineSavingsMsg'
Learn More

1H-Indazole-3-carboxylic Acid, Mikromol™

Product Code. 30050062
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050062 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050062 Supplier Mikromol Supplier No. MM0398.080025
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1H-Indazole-3-carboxylic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1H-Indazole-3-carboxylic Acid
CAS 4498-67-3
Analyte or Component Names 1H-Indazole-3-carboxylic Acid
Shipping Condition Room Temperature
Molecular Formula C8 H6 N2 O2
Certifications/Compliance ISO 17034
Synonym Granisetron Hydrochloride Imp. H (EP), Granisetron Imp. H (EP), 1H-Indazole-3-carboxylic Acid, Granisetron Hydrochloride Impurity H, Granisetron Impurity H
InChI Formula InChI=1S/C8H6N2O2/c11-8(12)7-5-3-1-2-4-6(5)9-10-7/h1-4H,(H,9,10)(H,11,12)
SMILES OC(=O)c1n[nH]c2ccccc12
IUPAC Name 1H-indazole-3-carboxylic acid
Molecular Weight (g/mol) 162.145
Quantity 25 mg
Formula Weight 162.043 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.