missing translation for 'onlineSavingsMsg'
Learn More
Learn More
d,l-Pheniramine.maleate, 1mg/ml in Methanol (as free base), Lipomed
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
d,l-Pheniramine.maleate, 1mg/ml in Methanol (as free base)
Specifications
Specifications
| Chemical Name or Material | Pheniramine Maleate |
| CAS | 132-20-7 |
| Analyte or Component Names | Pheniramine Maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H20 N2 . C4 H4 O4 |
| Concentration | 1 mg/mL |
| Synonym | Brompheniramine Maleate Imp. C (EP), Brompheniramine Imp. C (EP), Dexchlorpheniramine Maleate Imp. A (EP), Dexchlorpheniramine Imp. A (EP), (3 RS)-N,N-Dimethyl-3-phenyl-3-(pyridin-2-yl)propan-1-amine Maleate, N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1) |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CN(C)CCC(c1ccccc1)c2ccccn2.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;N,N-dimethyl-3-phenyl-3-pyridin-2-ylpropan-1-amine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction