missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mepivacaine, 1mg/ml in Methanol, Lipomed
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Mepivacaine, 1mg/ml in Methanol
Specifications
Specifications
| Chemical Name or Material | Mepivacaine |
| CAS | 96-88-8 |
| Analyte or Component Names | Mepivacaine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H22 N2 O |
| Concentration | 1 mg/mL |
| Synonym | 2-Piperidinecarboxamide, N-(2,6-dimethylphenyl)-1-methyl-, 2',6'-Pipecoloxylidide, 1-methyl- (6 CI,8 CI), N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide, (±)-Mepivacaine, 1-Methyl-2',6'-pipecoloxylidide, APF 135, Carbocain, Carbocaine, Carbocaine-V, DL-Mepivacaine, MepiSV, Mepicaine, Mepivacaine, Scandicain, Scandicaine, Tevacaine |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CN1CCCCC1C(=O)Nc2c(C)cccc2C |
| IUPAC Name | N-(2,6-dimethylphenyl)-1-methyl-piperidine-2-carboxamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction