missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylhexanamine.HCl, 1mg/ml in Methanol (as free base), Lipomed
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Methylhexanamine.HCl, 1mg/ml in Methanol (as free base)
Specifications
Specifications
| Chemical Name or Material | Methylhexanamine hydrochloride |
| CAS | 13803-74-2 |
| Analyte or Component Names | Methylhexanamine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H17 N . Cl H |
| Concentration | 1 mg/mL |
| Synonym | Dimethylamylamine Hydrochloride, 4-Methyl-2-hexanamine Hydrochloride, DMAA HCl |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.CCC(C)CC(C)N |
| IUPAC Name | 4-methylhexan-2-amine;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction