missing translation for 'onlineSavingsMsg'
Learn More

Naproxen, 1mg/ml in Methanol, Lipomed

Product Code. 30458194
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30458194

Brand: Lipomed LPMNPR17181LM

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Naproxen, 1mg/ml in Methanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Naproxen
CAS 22204-53-1
Analyte or Component Names Naproxen
Shipping Condition Room Temperature
Molecular Formula C14 H14 O3
Concentration 1 mg/mL
Synonym (+)-(S)-Naproxen, (+)-2-(6-Methoxy-2-naphthyl)propionic acid, (+)-Naproxen, (S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid, (S)-(+)-Naproxen, 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI), 2-Naphthaleneacetic acid, 6-methoxy-alpha-methyl-, (+)- (8 CI)
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES COc1ccc2cc(ccc2c1)[C@H](C)C(=O)O
IUPAC Name (2 S)-2-(6-methoxynaphthalen-2-yl)propanoic acid
Solvent or Matrix Methanol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 230.26
Quantity 1 mL
Formula Weight 230.0943 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.