missing translation for 'onlineSavingsMsg'
Learn More

Zopiclone Oxide 1.0 mg/ml in Dimethyl Sulfoxide, LoGiCal

Product Code. 30461701
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30461701

Brand: LoGiCal LGCAMP0149.0129

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Zopiclone Oxide 1.0 mg/ml in Dimethyl Sulfoxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zopiclone-N-oxide
CAS 43200-96-0
Analyte or Component Names Zopiclone-N-oxide
Shipping Condition Ice Pack (-20°C)
Molecular Formula C17 H17 Cl N6 O4
Certifications/Compliance ISO 17034
Synonym Zopiclone Imp. A (EP), Eszopiclone USP Related Compound A, Eszopiclone USP RC A, (5 RS)-6-(5-Chloropyridin-2-yl)-7-oxo-6,7-dihydro-5 H-pyrrolo[3,4-b]pyrazin-5-yl 4-Methylpiperazine-1-carboxylate 4-Oxide, Zopiclone Oxide, Eszopiclone Related Compound A, Zopiclone Impurity A
InChI Formula InChI=1 S/C17H17ClN6O4/c1-24(27)8-6-22(7-9-24)17(26)28-16-14-13(19-4-5-20-14)15(25)23(16)12-3-2-11(18)10-21-12/h2-5,10,16 H,6-9H2,1H3
SMILES C[N+]1([O-])CCN(CC1)C(=O)OC2N(C(=O)c3nccnc23)c4ccc(Cl)cn4
IUPAC Name [6-(5-chloropyridin-2-yl)-5-oxo-7 H-pyrrolo[3,4-b]pyrazin-7-yl] 4-methyl-4-oxidopiperazin-4-ium-1-carboxylate
Solvent or Matrix Dimethyl Sulfoxide
Industry Type Food Analysis
Molecular Weight (g/mol) 404.81
Quantity 1 mL
Formula Weight 404.1 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.