missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R,4S,5S,7s)-9-Butyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-7-yl (2S)-3-Hydroxy-2-phenylpropanoate Hydrochloride (N-Butylnorhyoscine Hydrochloride), Mikromol™

Product Code. 18022910
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18022910 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18022910 Supplier Mikromol Supplier No. MM0683.050025
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(1R,2R,4S,5S,7s)-9-Butyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-7-yl (2S)-3-Hydroxy-2-phenylpropanoate Hydrochloride (N-Butylnorhyoscine Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-butyl Norscopolamine hydrochloride
CAS 22235-98-9
Analyte or Component Names N-butyl Norscopolamine hydrochloride
Shipping Condition Room Temperature
Molecular Formula C20 H27 N O4 . Cl H
Certifications/Compliance ISO 17025
Synonym Benzeneacetic acid, α-(hydroxymethyl)-, 9-butyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl ester, hydrochloride, [7(S)-(1α,2β,4β,5α,7β)]- (9CI), 1αH,5αH-Nortropan-3α-ol, 8-butyl-6β,7β-epoxy-, (-)-tropate (ester), hydrochloride (8CI), 3-Oxa-9-azatricyclo[3.3.1.02,4]nonane, benzeneacetic acid deriv., Tropic acid, 8-butyl-6β,7β-epoxy-1αH,5αH-nortropan-3α-yl ester, hydrochloride, (-)- (8CI), N-Butylnorhyoscine Hydrochloride, N-Butylnorscopolamine Hydrochloride, (1R,2R,4S,5S,7s)-9-Butyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-7-yl (2S)-3-hydroxy-2-phenylpropanoate Hydrochloride, Hyoscine Butylbromide Imp. E (EP) as Hydrochloride, Hyoscine Butylbromide Impurity E as Hydrochloride
InChI Formula InChI=1S/C20H27NO4.ClH/c1-2-3-9-21-16-10-14(11-17(21)19-18(16)25-19)24-20(23)15(12-22)13-7-5-4-6-8-13;/h4-8,14-19,22H,2-3,9-12H2,1H3;1H/t14-,15-,16-,17+,18-,19+;/m1./s1
SMILES Cl.CCCCN1[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)[C@H](CO)c4ccccc4
Molecular Weight (g/mol) 381.89
Quantity 25 mg
Formula Weight 381.1707 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.