missing translation for 'onlineSavingsMsg'
Learn More

(1RS)-1-[[(Cyclohexyloxy)carbonyl]-oxy]ethyl 2-Oxo-3-[[2'-(1H-tetrazol-5-yl-)-biphenyl-4-yl]methyl]-2,3-dihydro-1H-benzimidazole-4-carboxylate, Mikromol™

Product Code. 30050884
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050884 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050884 Supplier Mikromol Supplier No. MM0776.020025
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(1RS)-1-[[(Cyclohexyloxy)carbonyl]-oxy]ethyl 2-Oxo-3-[[2'-(1H-tetrazol-5-yl-)-biphenyl-4-yl]methyl]-2,3-dihydro-1H-benzimidazole-4-carboxylate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O-Desethyl Candesartan Cilexetil
CAS 869631-11-8
Analyte or Component Names O-Desethyl Candesartan Cilexetil
Shipping Condition Room Temperature
Molecular Formula C31 H30 N6 O6
Certifications/Compliance ISO 9001
Synonym O-Desethyl Candesartan Cilexetil, (1RS)-1-[[(Cyclohexyloxy)carbonyl]-oxy]ethyl 2-Oxo-3-[[2'-(1H-tetrazol-5-yl-)-biphenyl-4-yl]methyl]-2,3-dihydro-1H-benzimidazole-4-carboxylate, 1H-Benzimidazole-4-carboxylic acid, 2,3-dihydro-2-oxo-3-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, 1H-Benzimidazole-4-carboxylic acid, 2,3-dihydro-2-oxo-3-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester (9CI), Candesartan Cilexetil Imp. B (EP)
InChI Formula InChI=1S/C31H30N6O6/c1-19(42-31(40)43-22-8-3-2-4-9-22)41-29(38)25-12-7-13-26-27(25)37(30(39)32-26)18-20-14-16-21(17-15-20)23-10-5-6-11-24(23)28-33-35-36-34-28/h5-7,10-17,19,22H,2-4,8-9,18H2,1H3,(H,32,39)(H,33,34,35,36)
SMILES CC(OC(=O)OC1CCCCC1)OC(=O)c2cccc3NC(=O)N(Cc4ccc(cc4)c5ccccc5c6nnn[nH]6)c23
IUPAC Name 1-cyclohexyloxycarbonyloxyethyl 2-oxo-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]-1H-benzimidazole-4-carboxylate
Molecular Weight (g/mol) 582.61
Quantity 25 mg
Formula Weight 582.2227 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.