missing translation for 'onlineSavingsMsg'
Learn More

(1RS,4SR)-7-(2-Methylpropyl)-1-[4-(2-methyl-propyl)phenyl]-1,2,3,4-tetrahydronaphthalene-1,4-dicarboxylic Acid, Mikromol™

Product Code. 30463641
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30463641 4 x 25 mg -
17984286 100 mg 100mg
17900996 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30463641 Supplier Mikromol Supplier No. MM0002.2900254AMP
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(1RS,4SR)-7-(2-Methylpropyl)-1-[4-(2-methyl-propyl)phenyl]-1,2,3,4-tetrahydronaphthalene-1,4-dicarboxylic Acid, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1RS,4SR)-7-(2-Methylpropyl)-1-[4-(2-methyl-propyl)phenyl]-1,2,3,4-tetrahydronaphthalene-1,4-dicarboxylic Acid
CAS 2517611-89-9
Analyte or Component Names (1RS,4SR)-7-(2-Methylpropyl)-1-[4-(2-methyl-propyl)phenyl]-1,2,3,4-tetrahydronaphthalene-1,4-dicarboxylic Acid
Shipping Condition Ice pack (-20°C)
Molecular Formula C26 H32 O4
Certifications/Compliance ISO 17025
Synonym (1RS,4SR)-7-(2-Methylpropyl)-1-[4-(2-methyl-propyl)phenyl]-1,2,3,4-tetrahydronaphthalene-1,4-dicarboxylic Acid
InChI Formula InChI=1S/C26H32O4/c1-16(2)13-18-5-8-20(9-6-18)26(25(29)30)12-11-22(24(27)28)21-10-7-19(14-17(3)4)15-23(21)26/h5-10,15-17,22H,11-14H2,1-4H3,(H,27,28)(H,29,30)/t22-,26+/m0/s1
Solution Type Neat
SMILES CC(C)Cc1ccc(cc1)[C@@]2(CC[C@H](C(=O)O)c3ccc(CC(C)C)cc23)C(=O)O
IUPAC Name (1S,4R)-6-(2-methylpropyl)-4-[4-(2-methylpropyl)phenyl]-2,3-dihydro-1H-naphthalene-1,4-dicarboxylic acid
Molecular Weight (g/mol) 408.53
Quantity 4 x 25 mg
Formula Weight 408.2301 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.