missing translation for 'onlineSavingsMsg'
Learn More

2-[1-methyl-3-(trifluoromethyl)-1h-pyrazol-5-yl]benzoic acid, 97%, Thermo Scientific™

Product Code. 10662295
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10662295 250 mg Amber Glass Bottle 250mg
10779595 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10662295 Supplier Thermo Scientific Maybridge Supplier No. CC62101CB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 910037-16-0
Molecular Formula C12H9F3N2O2
Molecular Weight (g/mol) 270.211
MDL Number MFCD09879926
InChI Key ADEAPHCAMGDLFU-UHFFFAOYSA-N
Synonym 2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl benzoic acid, 2-2-methyl-5-trifluoromethyl pyrazol-3-yl benzoic acid
PubChem CID 24229655
IUPAC Name 2-[2-methyl-5-(trifluoromethyl)pyrazol-3-yl]benzoic acid
SMILES CN1C(=CC(=N1)C(F)(F)F)C2=CC=CC=C2C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 270.21
Percent Purity 97%
Chemical Name or Material 2-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.