missing translation for 'onlineSavingsMsg'
Learn More

2-[2-[(2,6-Dichlorophenyl)amino]phenyl]acetic Acid (Diclofenac), Mikromol™

Product Code. 18073490
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18073490 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18073490 Supplier Mikromol Supplier No. MM0006.200250
 Restricted Item
Estimated Shipment: 15-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-[2-[(2,6-Dichlorophenyl)amino]phenyl]acetic Acid (Diclofenac)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diclofenac
CAS 15307-86-5
Analyte or Component Names Diclofenac
Shipping Condition Room Temperature
Molecular Formula C14 H11 Cl2 N O2
Certifications/Compliance ISO 17034
Synonym [2-[(2,6-Dichlorophenyl)amino]phenyl]acetic Acid, 2-[2-[(2,6-Dichlorophenyl)amino]phenyl]acetic Acid, Aceclofenac Imp. A (EP), Diclofenac, Acetic acid, [o-(2,6-dichloroanilino)phenyl]- (8CI), 2-(2,6-Dichloroanilino)phenylacetic acid, 2-(2,6-Dichlorophenylamino)phenylacetic acid, 2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid, Dichlofenac, Diclac, Diclofenac acid, Diclofenamic acid, Diclomelan, Dicloreuma, N-(2,6-Dichlorophenyl)-o-aminophenylacetic acid, Pennsaid, Transfenac, [o-(2,6-Dichloroanilino)phenyl]acetic acid, Aceclofenac Impurity A
InChI Formula InChI=1S/C14H11Cl2NO2/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19/h1-7,17H,8H2,(H,18,19)
SMILES OC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
IUPAC Name 2-[2-(2,6-dichloroanilino)phenyl]acetic acid
Molecular Weight (g/mol) 296.15
Quantity 250 mg
Formula Weight 295.0167 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.