missing translation for 'onlineSavingsMsg'
Learn More

2-(2,4-Difluorophenyl)propane-1,2,3-triol, Mikromol™

Product Code. 30050304
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050304 100 mg 100mg
30050628 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050304 Supplier Mikromol Supplier No. MM0235.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-(2,4-Difluorophenyl)propane-1,2,3-triol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2,4-Difluorophenyl)propane-1,2,3-triol
CAS 173837-65-5
Analyte or Component Names 2-(2,4-Difluorophenyl)propane-1,2,3-triol
Shipping Condition Room Temperature
Molecular Formula C9 H10 F2 O3
Certifications/Compliance ISO/IEC 17025
Synonym 2-(2,4-Difluorophenyl)propane-1,2,3-triol
InChI Formula InChI=1S/C9H10F2O3/c10-6-1-2-7(8(11)3-6)9(14,4-12)5-13/h1-3,12-14H,4-5H2
SMILES OCC(O)(CO)c1ccc(F)cc1F
IUPAC Name 2-(2,4-difluorophenyl)propane-1,2,3-triol
Molecular Weight (g/mol) 204.17
Quantity 100 mg
Formula Weight 204.0598 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.