missing translation for 'onlineSavingsMsg'
Learn More

[2-[(2-Amino-4-chlorophenyl)amino]phenyl](4-methyl-1-piperazinyl)methanone, TRC

Product Code. 30003396
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003396 100 mg 100mg
17994905 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003396 Supplier Toronto Research Chemicals Supplier No. TRCA603880100MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65514-71-8
Molecular Formula C18 H21 Cl N4 O
Molecular Weight (g/mol) 344.84
Synonym 1-[2-[(2-Amino-4-chlorophenyl)amino]benzoyl]-4-methylpiperazine, [2-[(2-Amino-4-chlorophenyl)amino]phenyl](4-methyl-1-piperazinyl)methanone
IUPAC Name [2-(2-amino-4-chloroanilino)phenyl]-(4-methylpiperazin-1-yl)methanone
SMILES CN1CCN(CC1)C(=O)c2ccccc2Nc3ccc(Cl)cc3N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H21ClN4O/c1-22-8-10-23(11-9-22)18(24)14-4-2-3-5-16(14)21-17-7-6-13(19)12-15(17)20/h2-7,12,21H,8-11,20H2,1H3
Formula Weight 344.1404
Chemical Name or Material 1-[2-[(2-Amino-4-chlorophenyl)amino]benzoyl]-4-methylpiperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.