CAS RN 65514-71-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1-[2-[(2-Amino-4-chlorophenyl)amino]benzoyl]-4-methylpiperazine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
[2-[(2-Amino-4-chlorophenyl)amino]phenyl](4-methyl-1-piperazinyl)methanone, TRC
CAS: 65514-71-8 Molecular Formula: C18 H21 Cl N4 O Molecular Weight (g/mol): 344.84 Synonym: 1-[2-[(2-Amino-4-chlorophenyl)amino]benzoyl]-4-methylpiperazine,[2-[(2-Amino-4-chlorophenyl)amino]phenyl](4-methyl-1-piperazinyl)methanone IUPAC Name: [2-(2-amino-4-chloroanilino)phenyl]-(4-methylpiperazin-1-yl)methanone SMILES: CN1CCN(CC1)C(=O)c2ccccc2Nc3ccc(Cl)cc3N