missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]propane-1,3-diyl Dipropanoate (Ganciclovir Dipropionate), Mikromol™
Description
2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]propane-1,3-diyl Dipropanoate (Ganciclovir Dipropionate)
Specifications
Specifications
| Chemical Name or Material | 2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]propane-1,3-diyl Dipropanoate (Ganciclovir Dipropionate) |
| CAS | 86357-20-2 |
| Analyte or Component Names | 2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]propane-1,3-diyl Dipropanoate (Ganciclovir Dipropionate) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H21 N5 O6 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ganciclovir Imp. I (EP), 2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]propane-1,3-diyl Dipropanoate, Ganciclovir Dipropionate, Ganciclovir Impurity I |
| InChI Formula | InChI=1S/C15H21N5O6/c1-3-10(21)24-5-9(6-25-11(22)4-2)26-8-20-7-17-12-13(20)18-15(16)19-14(12)23/h7,9H,3-6,8H2,1-2H3,(H3,16,18,19,23) |
| SMILES | CCC(=O)OCC(COC(=O)CC)OCn1cnc2C(=O)NC(=Nc12)N |
| IUPAC Name | [2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]-3-propanoyloxypropyl] propanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?