missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Carboxyvinyl)benzeneboronic acid, 98%, Thermo Scientific™

Product Code. 11309135
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309135 50 mg 50mg
11319135 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309135 Supplier Thermo Scientific Alfa Aesar Supplier No. L16368.MF
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 374105-86-9
Molecular Formula C9H9BO4
Molecular Weight (g/mol) 191.977
MDL Number MFCD02093067
InChI Key OPFDGNUJQALQFL-AATRIKPKSA-N
Synonym 2-2-carboxyvinyl benzeneboronic acid, 2-boronocinnamic acid, 3-2-boronophenyl acrylic acid, e-3-2-boronophenyl prop-2-enoic acid, e-3-2-boronophenyl acrylic acid, 2e-3-2-dihydroxyboranyl phenyl prop-2-enoic acid, 2-2-carboxyvinylbenzene boronic acid, 2-2-carboxyvinyl phenylboronic acid, 2-2-carboxy-vinyl-phenylboronic acid, e-3-2-boronophenyl-2-propenoic acid
PubChem CID 6158001
IUPAC Name (E)-3-(2-boronophenyl)prop-2-enoic acid
SMILES B(C1=CC=CC=C1C=CC(=O)O)(O)O

Specifications

Melting Point 170°C to 172°C
Quantity 50 mg
Formula Weight 191.98
Percent Purity 98%
Chemical Name or Material 2-(2-Carboxyvinyl)benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.