missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Iminodiacetic Acid (Iminodiacetic Acid), Mikromol™

Product Code. 30052226
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052226 100 mg 100mg
30052184 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30052226 Supplier Mikromol Supplier No. MM1509.01
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,2'-Iminodiacetic Acid (Iminodiacetic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iminodiacetic acid
CAS 142-73-4
Analyte or Component Names Iminodiacetic acid
Shipping Condition Room Temperature
Molecular Formula C4 H7 N O4
Certifications/Compliance ISO 17025
Synonym Glycine Imp. A (EP), N-(carboxymethyl)-Glycine, Acetic acid, iminodi- (6CI,8CI), ((Carboxymethyl)amino)acetic acid, Acetic acid, 2,2'-iminobis-, Aminodiacetic acid, Diglycin, Diglycine, Diglycocoll, IDA, IDA (chelating agent), IMDA, Iminobis(acetic acid), Iminodiacetic acid, Iminodiethanoic acid, N-(Carboxymethyl)glycine, NSC 18467, alpha-Iminodiacetic acid, Glycine Impurity A
InChI Formula InChI=1S/C4H7NO4/c6-3(7)1-5-2-4(8)9/h5H,1-2H2,(H,6,7)(H,8,9)
SMILES OC(=O)CNCC(=O)O
IUPAC Name 2-(carboxymethylamino)acetic acid
Molecular Weight (g/mol) 133.1
Quantity 100 mg
Formula Weight 133.0375 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.