missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Iminodiethanol (Diethanolamine), Mikromol™

Product Code. 17961526
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17961526 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17961526 Supplier Mikromol Supplier No. MM1204.020100
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,2'-Iminodiethanol (Diethanolamine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethanolamine
CAS 111-42-2
Analyte or Component Names Diethanolamine
Shipping Condition Room Temperature
Molecular Formula C4 H11 N O2
Certifications/Compliance ISO 9001
Synonym 2,2'-Iminobisethanol, Ethanol, 2,2'-iminodi- (8CI), 2,2'-Azanediyldiethanol, 2,2'-Dihydroxydiethylamine, 2,2'-Iminobis[ethanol], 2,2'-Iminodi-1-ethanol, 2,2'-Iminodiethanol, 2-[(2-Hydroxyethyl)amino]ethanol, Bis(2-hydroxyethyl)amine, Bis(hydroxyethyl)amine, DEA, DEA 80, DEAO-LF, DEOA LF, Dabco DEOA-LF, Di(2-hydroxyethyl)amine, Di(β-hydroxyethyl)amine, Diethanolamine, Diethanolamine 80, Diolamine, Iminodiethanol, N,N-Bis(2-hydroxyethyl)amine, N,N-Di(2-hydroxyethyl)amine, N,N-Diethanolamine, NSC 4959, Niax DEOA-LF, Tegoamin DEOA 85
InChI Formula InChI=1S/C4H11NO2/c6-3-1-5-2-4-7/h5-7H,1-4H2
SMILES OCCNCCO
IUPAC Name 2-(2-hydroxyethylamino)ethanol
Molecular Weight (g/mol) 105.14
Quantity 100 mg
Formula Weight 105.079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.