missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methyl-4-nitro-1H-imidazol-1-yl)ethyl Benzoate, Mikromol™

Product Code. 30050044
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050044 25 mg 25mg
30050863 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050044 Supplier Mikromol Supplier No. MM0278.040025
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-(2-Methyl-4-nitro-1H-imidazol-1-yl)ethyl Benzoate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2-Methyl-4-nitro-1H-imidazol-1-yl)ethyl Benzoate
CAS 87009-72-1
Analyte or Component Names 2-(2-Methyl-4-nitro-1H-imidazol-1-yl)ethyl Benzoate
Shipping Condition Room Temperature
Molecular Formula C13 H13 N3 O4
Certifications/Compliance ISO 9001
Synonym 2-(2-Methyl-4-nitro-1H-imidazol-1-yl)ethyl Benzoate
InChI Formula InChI=1S/C13H13N3O4/c1-10-14-12(16(18)19)9-15(10)7-8-20-13(17)11-5-3-2-4-6-11/h2-6,9H,7-8H2,1H3
SMILES Cc1nc(cn1CCOC(=O)c2ccccc2)[N+](=O)[O-]
IUPAC Name 2-(2-methyl-4-nitroimidazol-1-yl)ethyl benzoate
Molecular Weight (g/mol) 275.26
Quantity 25 mg
Formula Weight 275.0906 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.