missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(Trifluoromethyl)phenyl]ethylamine, 95%

Product Code. 15441729
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15441729 1 g 1g
15451729 5 g 5g
15461729 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15441729 Supplier Thermo Scientific Alfa Aesar Supplier No. H64492.03
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 774-18-5 | C9H10F3N | 189.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 774-18-5
Molecular Formula C9H10F3N
Molecular Weight (g/mol) 189.18
MDL Number MFCD00128232
InChI Key LIERORLYMWHXDL-UHFFFAOYSA-N
Synonym 2-2-trifluoromethyl phenyl ethanamine, o-trifluoromethyl phenethylamine, 2-2-trifluoromethyl phenyl ethan-1-amine, 2-o-trifluoromethylphenyl ethylamine, 2-2-trifluoromethylphenyl ethylamine, 2-trifluoromethyl phenethylamine, 2-2-trifluoromethyl-phenyl-ethylamine, 2-2-trifluoromethyl phenyl ethyl amine, 2-trifluoromethylphenylethanamine, benzeneethanamine,2-trifluoromethyl
PubChem CID 69883
IUPAC Name 2-[2-(trifluoromethyl)phenyl]ethanamine
SMILES NCCC1=CC=CC=C1C(F)(F)F

Specifications

Color Yellow
Quantity 1 g
UN Number UN2735
Beilstein 7202211
Formula Weight 189.18
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-[2-(Trifluoromethyl)phenyl]ethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.