missing translation for 'onlineSavingsMsg'
Learn More

2,3:4,5-Bis-O-(1-methylethylidene)-Beta-D-fructopyranose 1-[[(diethylamino)carbonyl]sulfamate], TRC

Product Code. 30010877
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
Unit Size:
100mg
1g
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010877 25 mg 25mg
30136828 1 g 1g
30136783 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010877 Supplier Toronto Research Chemicals Supplier No. TRCB49507525MG
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 876403-98-4
SMILES CCN(CC)C(=O)NS(=O)(=O)OC[C@@]12OC[C@H]3OC(C)(C)O[C@H]3[C@@H]1OC(C)(C)O2

Specifications

Analyte or Component Names 2,3:4,5-Bis-O-(1-methylethylidene)-beta-D-fructopyranose 1-(((diethylamino)carbonyl)sulfamate)
Shipping Condition Room Temperature
Molecular Formula C17H30N2O9S
Synonym Topiramate Impurity, N-(Diethylamino)carbonyl Topiramate
InChI Formula InChI=1S/C17H30N2O9S/c1-7-19(8-2)14(20)18-29(21,22)24-10-17-13(27-16(5,6)28-17)12-11(9-23-17)25-15(3,4)26-12/h11-13H,7-10H2,1-6H3,(H,18,20)/t11-,12-,13+,17+/m1/s1
Solution Type Neat
IUPAC Name [(1R,2S,6S,9R)-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[7.3.0.02,6]dodecan-6-yl]methyl N-(diethylcarbamoyl)sulfamate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 438.49
Quantity 25 mg
Formula Weight 438.1672
Percent Purity >95%
Chemical Name or Material 2,3:4,5-Bis-O-(1-methylethylidene)-beta-D-fructopyranose 1-(((diethylamino)carbonyl)sulfamate)
Grade HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.