missing translation for 'onlineSavingsMsg'
Learn More

2,3:4,5-Bis-O-(1-methylethylidene)-beta-D-fructopyranose, Mikromol™

Product Code. 17918924
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17918924 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17918924 Supplier Mikromol Supplier No. MM1348.010025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,3:4,5-Bis-O-(1-methylethylidene)-beta-D-fructopyranose
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3:4,5-Bis-O-(1-methylethylidene)-beta-D-fructopyranose
CAS 20880-92-6
Analyte or Component Names 2,3:4,5-Bis-O-(1-methylethylidene)-beta-D-fructopyranose
Shipping Condition Ice pack (-20°C)
Molecular Formula C12 H20 O6
Certifications/Compliance ISO 17025
Synonym 2,3:4,5-Bis-O-(1-methylethylidene)-beta-D-fructopyranose, Topiramate Related Compound A, Topiramate Impurity A, D-Fructopyranose Diacetonide, Diacetone Fructose, USP Topiramate Related Compound A
InChI Formula InChI=1S/C12H20O6/c1-10(2)15-7-5-14-12(6-13)9(8(7)16-10)17-11(3,4)18-12/h7-9,13H,5-6H2,1-4H3/t7-,8-,9+,12+/m1/s1
SMILES CC1(C)O[C@@H]2CO[C@@]3(CO)OC(C)(C)O[C@H]3[C@@H]2O1
IUPAC Name [(3aS,5aR,8aR,8bS)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydrodi[1,3]dioxolo[4,5-a:3',5'-c]pyran-3a-yl]methanol
Molecular Weight (g/mol) 260.28
Quantity 25 mg
Formula Weight 260.126 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.