missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[3-(4-Phenylpiperazin-1-yl)propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
2-[3-(4-Phenylpiperazin-1-yl)propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one
Specifications
Specifications
| Chemical Name or Material | 2-[3-(4-Phenylpiperazin-1-yl)propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one |
| CAS | 62337-66-0 |
| Analyte or Component Names | 2-[3-(4-Phenylpiperazin-1-yl)propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H23 N5 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-[3-(4-Phenylpiperazin-1-yl)propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one, Trazodone Hydrochloride Imp. B (BP), Trazodone Hydrochloride Impurity B, Trazodone Impurity B |
| InChI Formula | InChI=1S/C19H23N5O/c25-19-23-11-5-4-9-18(23)20-24(19)12-6-10-21-13-15-22(16-14-21)17-7-2-1-3-8-17/h1-5,7-9,11H,6,10,12-16H2 |
| SMILES | O=C1N(CCCN2CCN(CC2)c3ccccc3)N=C4C=CC=CN14 |
| IUPAC Name | 2-[3-(4-phenylpiperazin-1-yl)propyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction