missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrakis-O-(trimethylsilyl)Idono-Gamma-Lactone(>90%), TRC

Product Code. 30454750
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454750 100 mg 100mg
30449546 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30454750 Supplier Toronto Research Chemicals Supplier No. TRCT305830100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3,5,6-Tetrakis-O-(trimethylsilyl)Idono-Gamma-Lactone
Analyte or Component Names 2,3,5,6-Tetrakis-O-(trimethylsilyl)Idono-Gamma-Lactone
Shipping Condition Room Temperature
Molecular Formula C18 H42 O6 Si4
Synonym 2,3,5,6-Tetrakis-O-(trimethylsilyl)Idonic Acid gamma-Lactone, (3R,4S,5R)-5-((S)-2,2,7,7-Tetramethyl-3,6-dioxa-2,7-disilaoctan-4-yl)-3,4-bis((trimethylsilyl)oxy)dihydrofuran-2(3H)-one
InChI Formula InChI=1S/C18H42O6Si4/c1-25(2,3)20-13-14(22-26(4,5)6)15-16(23-27(7,8)9)17(18(19)21-15)24-28(10,11)12/h14-17H,13H2,1-12H3/t14-,15+,16-,17+/m0/s1
Solution Type Neat
SMILES C[Si](C)(C)OC[C@H](O[Si](C)(C)C)[C@H]1OC(=O)[C@H](O[Si](C)(C)C)[C@H]1O[Si](C)(C)C
IUPAC Name (3R,4S,5R)-5-[(1S)-1,2-bis(trimethylsilyloxy)ethyl]-3,4-bis(trimethylsilyloxy)oxolan-2-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 466.865
Quantity 100 mg
Formula Weight 466.206
Show More Show Less
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.