missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trifluorobenzyl bromide, 97%, Thermo Scientific™

Product Code. p-7047424
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374066 1 g 1g
11384066 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374066 Supplier Thermo Scientific Alfa Aesar Supplier No. L19191.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 226717-83-5
Molecular Formula C7H4BrF3
Molecular Weight (g/mol) 225.008
MDL Number MFCD00083543
InChI Key QPMAEHBAXJSICW-UHFFFAOYSA-N
Synonym 2,3,5-trifluorobenzyl bromide, 1-bromomethyl-2,3,5-trifluorobenzene, 2,3,5-trifluorobenzylbromide, benzene, 1-bromomethyl-2,3,5-trifluoro, acmc-1chwx, 2,3,5-trifluoro benzyl bromide, 1-bromomethyl-2,3,5-trifluoro-benzene, alpha-bromo-2,3,5-trifluorotoluene, benzene,1-bromomethyl-2,3,5-trifluoro, alpha-bromo-2,3,5-trifluorotoluene;a-bromo-2,3,5-trifluorotoluene
PubChem CID 2777052
IUPAC Name 1-(bromomethyl)-2,3,5-trifluorobenzene
SMILES C1=C(C=C(C(=C1F)F)CBr)F

Specifications

Refractive Index 1.505
Quantity 1 g
UN Number UN3265
Beilstein 8311831
Formula Weight 225.01
Percent Purity 97%
Chemical Name or Material 2,3,5-Trifluorobenzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.