missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzoic Acid, Mikromol™

Product Code. 17950796
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17950796 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17950796 Supplier Mikromol Supplier No. MM0922.050025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,3-Dichlorobenzoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3-Dichlorobenzoic acid
CAS 50-45-3
Analyte or Component Names 2,3-Dichlorobenzoic acid
Shipping Condition Room Temperature
Molecular Formula C7 H4 Cl2 O2
Certifications/Compliance ISO 17025
Synonym Lamotrigine Imp. E (EP), Lamotrigine USP Related Compound B, Lamotrigine USP RC B, 2,3-Dichlorobenzoic Acid, USP Lamotrigine Related Compound B, 69Y77, Ph Eur Lamotrigine Impurity E, 2,3-dichlorobenzoic acid 69Y77 (Ph Eur Lamotrigine Impurity E), (Ph Eur Impurity E), Lamotrigine Related Compound B, Lamotrigine Impurity E
InChI Formula InChI=1S/C7H4Cl2O2/c8-5-3-1-2-4(6(5)9)7(10)11/h1-3H,(H,10,11)
SMILES OC(=O)c1cccc(Cl)c1Cl
IUPAC Name 2,3-dichlorobenzoic acid
Molecular Weight (g/mol) 191.01
Quantity 25 mg
Formula Weight 189.9588 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.