missing translation for 'onlineSavingsMsg'
Learn More

2-[4-[3-[5-Oxo-2-(trifluoromethyl)-10H-5lambda4-phenothiazin-10-yl]propyl]piperazin-1-yl]ethanol (Fluphenazine S-Oxide), Mikromol™

Product Code. 30050741
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050741 25 mg 25mg
30049131 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050741 Supplier Mikromol Supplier No. MM0114.010025
 Restricted Item
Estimated Shipment: 15-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-[4-[3-[5-Oxo-2-(trifluoromethyl)-10H-5lambda4-phenothiazin-10-yl]propyl]piperazin-1-yl]ethanol (Fluphenazine S-Oxide)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluphenazine S-Oxide
CAS 1674-76-6
Analyte or Component Names Fluphenazine S-Oxide
Shipping Condition Room Temperature
Molecular Formula C22 H26 F3 N3 O2 S
Certifications/Compliance ISO 17025
Synonym Fluphenazine Dihydrochloride Imp. A (EP), Fluphenazine Imp. A (EP), 2-[4-[3-[5-Oxo-2-(trifluoromethyl)-10H-5lambda4-phenothiazin-10-yl]propyl]piperazin-1-yl]ethanol, Fluphenazine S-Oxide, Fluphenazine Related Compound A, Fluphenazine Dihydrochloride Impurity A, Fluphenazine Impurity A
InChI Formula InChI=1S/C22H26F3N3O2S/c23-22(24,25)17-6-7-21-19(16-17)28(18-4-1-2-5-20(18)31(21)30)9-3-8-26-10-12-27(13-11-26)14-15-29/h1-2,4-7,16,29H,3,8-15H2
SMILES OCCN1CCN(CCCN2c3ccccc3S(=O)c4ccc(cc24)C(F)(F)F)CC1
IUPAC Name 2-[4-[3-[5-oxo-2-(trifluoromethyl)phenothiazin-10-yl]propyl]piperazin-1-yl]ethanol
Molecular Weight (g/mol) 453.52
Quantity 25 mg
Formula Weight 453.1698 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.